Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrpl10
Entrez Gene ID
691075
Gene Name
mitochondrial ribosomal protein L10
Gene Aliases
L10mt, MRP-L10, MRP-L8
Chromosome Location
Chr.10: 84976377 - 84984793 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109620.1 NP_001103090.1 2 - 3 269 124

View Full Product Details