Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnk1
Entrez Gene ID
303247
Gene Name
tyrosine kinase, non-receptor, 1
Gene Aliases
-
Chromosome Location
Chr.10: 56458650 - 56468449 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107012.1 NP_001100482.1 9 - 10 1532 81
XM_006246704.2 XP_006246766.1 10 - 11 2990 81
XM_006246705.2 XP_006246767.1 11 - 12 2065 81
XM_006246707.2 XP_006246769.1 11 - 12 1735 81
XM_006246709.2 XP_006246771.1 10 - 11 1912 81
XM_008767827.1 XP_008766049.1 10 - 11 2441 81
XM_008767828.1 XP_008766050.1 11 - 12 1829 81

View Full Product Details