The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181086.3 | NP_851600.1 | 3 - 4 | 364 | 63 |
GenBank mRNA | AY255102.1 | - | 3 - 4 | 335 | 63 |
BC060537.1 | - | 3 - 4 | 368 | 63 | |
FQ229590.1 | - | 3 - 4 | 364 | 63 |