The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108299.1 | NP_001101769.1 | 6 - 7 | 657 | 66 |
XM_006247369.2 | XP_006247431.1 | 6 - 7 | 730 | 66 | |
XM_006247370.2 | XP_006247432.1 | 6 - 7 | 678 | 66 | |
XM_006247371.2 | XP_006247433.1 | 6 - 7 | 638 | 66 | |
XM_006247372.2 | XP_006247434.1 | 6 - 7 | 485 | 66 | |
XM_008768085.1 | XP_008766307.1 | 6 - 7 | 727 | 66 | |
GenBank mRNA | BC168950.1 | - | 6 - 7 | 531 | 66 |