The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107060.1 | NP_001100530.1 | 5 - 6 | 1422 | 110 |
GenBank mRNA | BC091245.1 | - | 5 - 6 | 943 | 110 |