Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcaf7
Entrez Gene ID
303602
Gene Name
DDB1 and CUL4 associated factor 7
Gene Aliases
RGD1305140, Wdr68
Chromosome Location
Chr.10: 94231585 - 94254046 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107057.3 NP_001100527.1 4 - 5 749 61
GenBank mRNA BC167045.1 - 4 - 5 749 61

View Full Product Details