Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nags
Entrez Gene ID
303563
Gene Name
N-acetylglutamate synthase
Gene Aliases
RGD1565783
Chromosome Location
Chr.10: 90084609 - 90089693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107053.1 NP_001100523.1 6 - 7 1500 126
XM_006247328.2 XP_006247390.1 7 - 8 2320 126
XM_008768055.1 XP_008766277.1 7 - 8 2320 126

View Full Product Details