The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133418.1 | NP_596909.1 | 10 - 11 | 799 | 88 |
GenBank mRNA | AJ223355.1 | - | 10 - 11 | 799 | 88 |
BC081734.1 | - | 10 - 11 | 817 | 88 |