Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rilp
Entrez Gene ID
287531
Gene Name
Rab interacting lysosomal protein
Gene Aliases
-
Chromosome Location
Chr.10: 63659030 - 63662188 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105811.1 NP_001099281.1 5 - 6 805 85

View Full Product Details