Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Diaph3
Entrez Gene ID
290396
Gene Name
diaphanous-related formin 3
Gene Aliases
Diap3
Chromosome Location
Chr.15: 69928507 - 70400055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728299.4 XP_002728345.2 7 - 8 838 87

View Full Product Details