The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014139.1 | NP_001014161.1 | 2 - 3 | 364 | 61 |
GenBank mRNA | BC088867.1 | - | 2 - 3 | 364 | 61 |