Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tax1bp3
Entrez Gene ID
360564
Gene Name
Tax1 (human T-cell leukemia virus type I) binding protein 3
Gene Aliases
-
Chromosome Location
Chr.10: 59743356 - 59748063 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246761.2 XP_006246823.1 2 - 3 399 76
GenBank mRNA FQ215653.1 - 2 - 3 211 76
FQ219871.1 - 2 - 3 76
FQ220030.1 - 2 - 3 212 76
FQ220071.1 - 2 - 3 211 76
FQ220113.1 - 2 - 3 211 76
FQ229275.1 - 2 - 3 211 76

View Full Product Details