Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dhrs7b
Entrez Gene ID
287380
Gene Name
dehydrogenase/reductase (SDR family) member 7B
Gene Aliases
RGD1311243
Chromosome Location
Chr.10: 47065750 - 47111089 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006246474.2 XP_006246536.1 1 - 2 364 63

View Full Product Details