Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl7
Entrez Gene ID
287561
Gene Name
chemokine (C-C motif) ligand 7
Gene Aliases
-
Chromosome Location
Chr.10: 69423083 - 69424933 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007612.1 NP_001007613.1 1 - 2 165 77

View Full Product Details