Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ypel2
Entrez Gene ID
360590
Gene Name
yippee-like 2 (Drosophila)
Gene Aliases
RGD1311564
Chromosome Location
Chr.10: 74241032 - 74298599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108286.1 NP_001101756.1 1 - 2 145 161
XM_006247097.1 XP_006247159.1 1 - 2 130 161

View Full Product Details