Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sbp
Entrez Gene ID
25540
Gene Name
spermine binding protein
Gene Aliases
-
Chromosome Location
Chr.10: 13289514 - 13309332 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013024.2 NP_037156.2 2 - 3 260 99
XM_008767548.1 XP_008765770.1 1 - 2 66 99
GenBank mRNA CA339008.1 - 2 - 3 260 99

View Full Product Details