Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim11
Entrez Gene ID
360534
Gene Name
tripartite motif-containing 11
Gene Aliases
-
Chromosome Location
Chr.10: 45322199 - 45335030 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108276.1 NP_001101746.1 5 - 6 994 90
XM_006246505.2 XP_006246567.1 5 - 6 993 90
GenBank mRNA BC160894.1 - 5 - 6 944 90

View Full Product Details