Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apoh
Entrez Gene ID
287774
Gene Name
apolipoprotein H (beta-2-glycoprotein I)
Gene Aliases
-
Chromosome Location
Chr.10: 96640013 - 96653939 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009626.1 NP_001009626.1 2 - 3 245 145
GenBank mRNA AM071386.1 - 2 - 3 204 145
BC088180.1 - 2 - 3 245 145
FQ209412.1 - 2 - 3 284 145
FQ209491.1 - 2 - 3 271 145
FQ209740.1 - 2 - 3 280 145
FQ210182.1 - 2 - 3 271 145
FQ210311.1 - 2 - 3 271 145
FQ210756.1 - 2 - 3 287 145
FQ210813.1 - 2 - 3 271 145
FQ211328.1 - 2 - 3 244 145
FQ218445.1 - 2 - 3 285 145
FQ218454.1 - 2 - 3 271 145
FQ218597.1 - 2 - 3 267 145
FQ218599.1 - 2 - 3 271 145
FQ218637.1 - 2 - 3 271 145
FQ218785.1 - 2 - 3 287 145
FQ218859.1 - 2 - 3 244 145
FQ218865.1 - 2 - 3 271 145
FQ218961.1 - 2 - 3 271 145
FQ219138.1 - 2 - 3 336 145
FQ219328.1 - 2 - 3 271 145
FQ219380.1 - 2 - 3 272 145
FQ219438.1 - 2 - 3 270 145
FQ219481.1 - 2 - 3 284 145
FQ219537.1 - 2 - 3 268 145
FQ219580.1 - 2 - 3 285 145
FQ219601.1 - 2 - 3 284 145
FQ219674.1 - 2 - 3 271 145

View Full Product Details