Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Limd2
Entrez Gene ID
360646
Gene Name
LIM domain containing 2
Gene Aliases
RGD1309967
Chromosome Location
Chr.10: 94350787 - 94353293 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247622.2 XP_006247684.1 1 - 2 179 92

View Full Product Details