Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sugt1
Entrez Gene ID
290408
Gene Name
SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae)
Gene Aliases
Aa1114
Chromosome Location
Chr.15: 61873158 - 61913983 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013051.1 NP_001013069.1 2 - 3 154 93
GenBank mRNA BC158724.1 - 2 - 3 154 93
FQ220550.1 - 2 - 3 189 93
FQ221379.1 - 2 - 3 180 93
FQ225182.1 - 2 - 3 272 93
FQ227975.1 - 2 - 3 272 93
FQ232174.1 - 2 - 3 272 93

View Full Product Details