Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sugt1
Entrez Gene ID
290408
Gene Name
SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae)
Gene Aliases
Aa1114
Chromosome Location
Chr.15: 61873158 - 61913983 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013051.1 NP_001013069.1 8 - 9 484 147
GenBank mRNA BC158724.1 - 8 - 9 484 147
FQ220550.1 - 8 - 9 519 147
FQ221379.1 - 8 - 9 510 147
FQ225182.1 - 8 - 9 602 147
FQ227975.1 - 8 - 9 602 147
FQ232174.1 - 8 - 9 602 147

View Full Product Details