Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elf1
Entrez Gene ID
85424
Gene Name
E74-like factor 1
Gene Aliases
-
Chromosome Location
Chr.15: 61772626 - 61868442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053520.2 NP_445972.2 2 - 3 486 146
XM_006252347.2 XP_006252409.1 3 - 4 1745 146
XM_006252348.2 XP_006252410.1 2 - 3 483 146
XM_006252349.2 XP_006252411.1 4 - 5 1911 146
XM_006252351.2 XP_006252413.1 3 - 4 989 146
XM_006252352.2 XP_006252414.1 3 - 4 949 146
XM_008770898.1 XP_008769120.1 3 - 4 3249 146
XM_008770899.1 XP_008769121.1 3 - 4 945 146
XM_008770900.1 XP_008769122.1 3 - 4 501 146
XM_008770901.1 XP_008769123.1 3 - 4 3249 146
GenBank mRNA AB046531.1 - 2 - 3 265 146
FQ221456.1 - 4 - 5 1054 146

View Full Product Details