Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elf1
Entrez Gene ID
85424
Gene Name
E74-like factor 1
Gene Aliases
-
Chromosome Location
Chr.15: 61772626 - 61868442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CB782606.1 - 1 - 2 54 88

View Full Product Details