The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107275.1 | NP_001100745.1 | 8 - 9 | 1371 | 66 |
XM_006252270.2 | XP_006252332.1 | 8 - 9 | 1396 | 66 |