Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rdh14
Entrez Gene ID
500629
Gene Name
retinol dehydrogenase 14 (all-trans/9-cis/11-cis)
Gene Aliases
RGD1565196
Chromosome Location
Chr.6: 36106926 - 36112069 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109276.1 NP_001102746.1 1 - 2 839 89

View Full Product Details