Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mapre3
Entrez Gene ID
298848
Gene Name
microtubule-associated protein, RP/EB family, member 3
Gene Aliases
-
Chromosome Location
Chr.6: 26878738 - 26923420 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764512.1 XP_008762734.1 1 - 2 149 156
XM_008764513.1 XP_008762735.1 1 - 2 149 156

View Full Product Details