Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp6v1d
Entrez Gene ID
299159
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit D
Gene Aliases
-
Chromosome Location
Chr.6: 102032271 - 102047754 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199386.2 NP_955418.1 6 - 7 532 108
GenBank mRNA BC063177.1 - 6 - 7 536 108
FQ213312.1 - 6 - 7 558 108
FQ214434.1 - 6 - 7 521 108
FQ216046.1 - 3 - 4 179 108

View Full Product Details