Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcfd2
Entrez Gene ID
246117
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
Sdnsf
Chromosome Location
Chr.6: 10887303 - 10899221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139253.1 NP_640346.1 3 - 4 390 72
XM_006239710.2 XP_006239772.1 3 - 4 429 72
XM_006239711.2 XP_006239773.1 3 - 4 450 72
GenBank mRNA AF475282.1 - 3 - 4 390 72
BC071179.1 - 3 - 4 384 72

View Full Product Details