Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcfd2
Entrez Gene ID
246117
Gene Name
multiple coagulation factor deficiency 2
Gene Aliases
Sdnsf
Chromosome Location
Chr.6: 10887303 - 10899221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239710.2 XP_006239772.1 1 - 2 106 82
GenBank mRNA BC071179.1 - 1 - 2 61 82

View Full Product Details