The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106710.1 | NP_001100180.1 | 7 - 8 | 3185 | 93 |
XM_006239793.2 | XP_006239855.1 | 7 - 8 | 3196 | 93 | |
XM_008764511.1 | XP_008762733.1 | 7 - 8 | 3928 | 93 |