Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eapp
Entrez Gene ID
299043
Gene Name
E2F-associated phosphoprotein
Gene Aliases
RGD1309624
Chromosome Location
Chr.6: 75561898 - 75589273 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106729.1 NP_001100199.1 2 - 3 348 69
NM_001134987.1 NP_001128459.1 2 - 3 332 69
XM_006240100.2 XP_006240162.1 2 - 3 373 69
GenBank mRNA BC168880.1 - 2 - 3 332 69

View Full Product Details