Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem30b
Entrez Gene ID
314224
Gene Name
transmembrane protein 30B
Gene Aliases
-
Chromosome Location
Chr.6: 96439607 - 96442682 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080380.1 NP_001073849.1 1 - 1 1333 66

View Full Product Details