Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bzw2
Entrez Gene ID
171439
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
Hfb2
Chromosome Location
Chr.6: 55586754 - 55647650 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134402.3 NP_599229.1 8 - 9 962 84
GenBank mRNA AF031483.1 - 8 - 9 990 84
BC063149.1 - 8 - 9 966 84
FQ225034.1 - 8 - 9 938 84
FQ228189.1 - 8 - 9 939 84

View Full Product Details