Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lgals8
Entrez Gene ID
116641
Gene Name
lectin, galactoside-binding, soluble, 8
Gene Aliases
-
Chromosome Location
Chr.17: 66487239 - 66515306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053862.2 NP_446314.2 7 - 8 689 98
XM_006254166.2 XP_006254228.1 7 - 8 955 98
XM_006254167.2 XP_006254229.1 7 - 8 701 98
XM_006254168.1 XP_006254230.1 7 - 8 728 98
GenBank mRNA BC072488.1 - 7 - 8 689 98
U09824.1 - 6 - 7 669 98

View Full Product Details