Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc16a13
Entrez Gene ID
287451
Gene Name
solute carrier family 16, member 13
Gene Aliases
-
Chromosome Location
Chr.10: 56827593 - 56831243 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005530.1 NP_001005530.1 1 - 2 485 137
GenBank mRNA BC081902.1 - 1 - 2 485 137

View Full Product Details