Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt34
Entrez Gene ID
303528
Gene Name
keratin 34
Gene Aliases
Ka29, Krt1-4
Chromosome Location
Chr.10: 87950364 - 87954055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008758.1 NP_001008758.1 2 - 3 429 58

View Full Product Details