Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Endov
Entrez Gene ID
498029
Gene Name
endonuclease V
Gene Aliases
-
Chromosome Location
Chr.10: 108630763 - 108646679 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC158682.1 - 2 - 3 233 56

View Full Product Details