The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108278.1 | NP_001101748.1 | 5 - 6 | 497 | 70 |
XM_006246743.1 | XP_006246805.1 | 6 - 7 | 618 | 70 | |
XM_006246744.2 | XP_006246806.1 | 6 - 7 | 519 | 70 | |
XM_008767837.1 | XP_008766059.1 | 5 - 6 | 534 | 70 | |
GenBank mRNA | FQ218736.1 | - | 6 - 7 | 478 | 70 |