Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cgref1
Entrez Gene ID
245918
Gene Name
cell growth regulator with EF hand domain 1
Gene Aliases
Cgr11
Chromosome Location
Chr.6: 26797126 - 26809132 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139087.2 NP_620787.2 4 - 5 359 69
GenBank mRNA FM040765.1 - 4 - 5 359 69
FQ220335.1 - 4 - 5 289 69
U66470.1 - 4 - 5 280 69

View Full Product Details