Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jkamp
Entrez Gene ID
299127
Gene Name
JNK1/MAPK8-associated membrane protein
Gene Aliases
RGD1308917
Chromosome Location
Chr.6: 94835845 - 94849784 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106738.1 NP_001100208.1 1 - 2 344 69

View Full Product Details