Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmx1
Entrez Gene ID
362751
Gene Name
thioredoxin-related transmembrane protein 1
Gene Aliases
Txndc1
Chromosome Location
Chr.6: 92864123 - 92875141 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024800.1 NP_001019971.1 5 - 6 526 94
XM_006240178.2 XP_006240240.1 5 - 6 1749 94
GenBank mRNA BC094308.1 - 5 - 6 526 94

View Full Product Details