Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Timmdc1
Entrez Gene ID
303922
Gene Name
translocase of inner mitochondrial membrane domain containing 1
Gene Aliases
RGD1359380
Chromosome Location
Chr.11: 64790801 - 64815484 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007676.1 NP_001007677.1 6 - 7 807 100
GenBank mRNA BC079140.1 - 6 - 7 807 100

View Full Product Details