The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012926.1 | NP_037058.1 | 4 - 5 | 879 | 74 |
XM_006248365.2 | XP_006248427.1 | 4 - 5 | 1719 | 74 | |
GenBank mRNA | AF010465.1 | - | 4 - 5 | 879 | 74 |
U05593.1 | - | 4 - 5 | 890 | 74 |