The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008337.1 | NP_001008338.1 | 1 - 2 | 52 | 90 |
GenBank mRNA | BC085895.1 | - | 1 - 2 | 52 | 90 |