Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb11
Entrez Gene ID
304010
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
-
Chromosome Location
Chr.11: 47079201 - 47113993 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107097.1 NP_001100567.1 1 - 2 474 88

View Full Product Details