Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a14
Entrez Gene ID
85263
Gene Name
solute carrier family 25 (mitochondrial carrier, brain), member 14
Gene Aliases
Bmcp1
Chromosome Location
Chr.X: 135470919 - 135509223 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773531.1 XP_008771753.1 1 - 2 68 91

View Full Product Details