Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhox4g
Entrez Gene ID
313451
Gene Name
reproductive homeobox 4G
Gene Aliases
-
Chromosome Location
Chr.X: 123851846 - 123857609 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024889.1 NP_001020060.1 2 - 3 516 65
GenBank mRNA DQ058652.1 - 2 - 3 516 65

Gene Symbol
LOC103689929
Entrez Gene ID
103689929
Gene Name
homeobox protein Rhox5-like
Gene Aliases
-
Chromosome Location
Chr.X: 124561214 - 124566977 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773509.1 XP_008771731.1 2 - 3 516 65

View Full Product Details