The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127504.1 | NP_001120976.1 | 5 - 6 | 712 | 68 |
XM_006229270.2 | XP_006229332.1 | 6 - 7 | 1177 | 68 | |
XM_006229271.2 | XP_006229333.1 | 5 - 6 | 631 | 68 | |
XM_006229273.2 | XP_006229335.1 | 6 - 7 | 969 | 68 | |
XM_006229274.2 | XP_006229336.1 | 6 - 7 | 1496 | 68 | |
XM_008759407.1 | XP_008757629.1 | 5 - 6 | 710 | 68 | |
XM_008759408.1 | XP_008757630.1 | 5 - 6 | 631 | 68 |