Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hrc
Entrez Gene ID
292905
Gene Name
histidine rich calcium binding protein
Gene Aliases
-
Chromosome Location
Chr.1: 101324788 - 101328391 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181369.2 NP_852034.1 5 - 6 2304 92
GenBank mRNA AY277279.1 - 5 - 6 2304 92
FQ216289.1 - 5 - 6 2398 92
FQ224148.1 - 5 - 6 2390 92

View Full Product Details