The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014810.1 | NP_001014810.1 | 7 - 8 | 812 | 86 |
NM_198734.2 | NP_942029.2 | 7 - 8 | 822 | 86 | |
XM_008759128.1 | XP_008757350.1 | 7 - 8 | 841 | 86 | |
XM_008759129.1 | XP_008757351.1 | 7 - 8 | 821 | 86 | |
GenBank mRNA | BC058153.1 | - | 6 - 7 | 659 | 86 |
CB809766.1 | - | 2 - 3 | 156 | 86 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006228658.2 | XP_006228720.1 | 7 - 8 | 841 | 86 |
XM_006228660.2 | XP_006228722.1 | 7 - 8 | 821 | 86 | |
XM_008759221.1 | XP_008757443.1 | 7 - 8 | 851 | 86 | |
XM_008759222.1 | XP_008757444.1 | 7 - 8 | 870 | 86 |